siRNA Details
Gene Symbol: Ptpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001059757.4 | XP_001059757.1 | 9 | 1275 | ||
XM_006238190.2 | XP_006238252.1 | 9 | 1048 | |||
XM_006238191.2 | XP_006238253.1 | 9 | 977 | |||
XM_006238192.2 | XP_006238254.1 | 9 | 746 | |||
XM_006238193.2 | XP_006238255.1 | 9 | 2355 | |||
XM_008763738.1 | XP_008761960.1 | 8 | 1230 | |||
XM_008763739.1 | XP_008761961.1 | 10 | 851 | |||
XM_008763740.1 | XP_008761962.1 | 2 | 97 |

More Information
Other Species Targeted

Gene Ontology Categories:
