siRNA Details
Gene Symbol: Ptpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001059757.4 | XP_001059757.1 | 25 | 3274 | ||
XM_006238190.2 | XP_006238252.1 | 25 | 3047 | |||
XM_006238191.2 | XP_006238253.1 | 25 | 2976 | |||
XM_006238192.2 | XP_006238254.1 | 25 | 2745 | |||
XM_006238193.2 | XP_006238255.1 | 25 | 4354 | |||
XM_008763738.1 | XP_008761960.1 | 24 | 3229 | |||
XM_008763739.1 | XP_008761961.1 | 26 | 2850 | |||
XM_008763740.1 | XP_008761962.1 | 18 | 2096 | |||
GenBank mRNA | BC092605.1 | - | 7 | 797 |

More Information
Other Species Targeted

Gene Ontology Categories:
