siRNA Details
Gene Symbol: Ptpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001059757.4 | XP_001059757.1 | 10 | 1384 | ||
XM_006238190.2 | XP_006238252.1 | 10 | 1157 | |||
XM_006238191.2 | XP_006238253.1 | 10 | 1086 | |||
XM_006238192.2 | XP_006238254.1 | 10 | 855 | |||
XM_006238193.2 | XP_006238255.1 | 10 | 2464 | |||
XM_008763738.1 | XP_008761960.1 | 9 | 1339 | |||
XM_008763739.1 | XP_008761961.1 | 11 | 960 | |||
XM_008763740.1 | XP_008761962.1 | 3 | 206 |
