siRNA Details
Gene Symbol: Lurap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025022.1 | NP_001020193.1 | 2 | 692 | ||
GenBank mRNA | BC091300.1 | - | 2 | 692 | 7309480 |

More Information
Other Species Targeted

