siRNA Details
Gene Symbol: Snapc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013212.1 | NP_001013230.1 | 1 | 186 | ||
GenBank mRNA | BC091186.1 | - | 1 | 186 | 7366548 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Snapc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013212.1 | NP_001013230.1 | 1 | 186 | ||
GenBank mRNA | BC091186.1 | - | 1 | 186 | 7366548 |