siRNA Details
Gene Symbol: Tctex1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135030.1 | NP_001128502.1 | 5 | 682 | ||
XM_006238459.2 | XP_006238521.1 | 5 | 1438 | |||
XM_008763888.1 | XP_008762110.1 | 4 | 509 | |||
XM_008763889.1 | XP_008762111.1 | 5 | 3355 | |||
XM_008763890.1 | XP_008762112.1 | 4 | 1216 | |||
XM_008763891.1 | XP_008762113.1 | 4 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
