siRNA Details
Gene Symbol: RGD1305347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108674.1 | NP_001102144.1 | 3 | 1020 | ||
XM_006238692.2 | XP_006238754.1 | 3 | 916 | |||
XM_006238693.2 | XP_006238755.1 | 4 | 1311 |

More Information
Other Species Targeted

