siRNA Details
Gene Symbol: Ccdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038992.1 | NP_001034081.1 | 2 | 192 | ||
NM_001038994.2 | NP_001034083.1 | 2 | 292 | |||
GenBank mRNA | BC099226.1 | - | 2 | 192 | 7388643 | |
BF559487.1 | - | 2 | 298 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
