siRNA Details
Gene Symbol: Zmym6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108681.2 | NP_001102151.1 | 12 | 1871 | ||
XM_006238892.2 | XP_006238954.1 | 11 | 2702 | |||
XM_006238894.2 | XP_006238956.1 | 12 | 1749 | |||
GenBank mRNA | FQ221018.1 | - | 11 | 2237 |
