siRNA Details
Gene Symbol: Tlr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108682.1 | NP_001102152.1 | 1 | 1387 | ||
XM_008764148.1 | XP_008762370.1 | 1 | 1941 | |||
XM_008764149.1 | XP_008762371.1 | 1 | 1941 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
