siRNA Details
Gene Symbol: Tlr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108682.1 | NP_001102152.1 | 1 | 2416 | ||
XM_008764148.1 | XP_008762370.1 | 1 | 2970 | |||
XM_008764149.1 | XP_008762371.1 | 2 | 2679 |

More Information
Other Species Targeted

Gene Ontology Categories:
