siRNA Details
Gene Symbol: Kpna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015029.2 | NP_001015029.2 | 1 | 89 | ||
GenBank mRNA | AY779030.1 | - | 1 | 39 | ||
BC105813.1 | - | 1 | 89 | 7112648 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008468, AF020773, AK154882, AK084212, CJ082540, and others.
Gene Ontology Categories:
