siRNA Details
Gene Symbol: Pum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108684.1 | NP_001102154.1 | 5 | 788 | ||
XM_006239029.1 | XP_006239091.1 | 5 | 815 | |||
XM_006239030.2 | XP_006239092.1 | 5 | 813 | |||
XM_006239031.1 | XP_006239093.1 | 5 | 815 | |||
XM_006239032.1 | XP_006239094.1 | 5 | 815 | |||
XM_006239033.1 | XP_006239095.1 | 5 | 815 | |||
XM_006239034.1 | XP_006239096.1 | 5 | 815 | |||
XM_006239035.1 | XP_006239097.1 | 5 | 815 | |||
XM_006239036.1 | XP_006239098.1 | 5 | 815 | |||
XM_008764150.1 | XP_008762372.1 | 5 | 789 |

More Information
Other Species Targeted

Gene Ontology Categories:
