siRNA Details
Gene Symbol: Cep85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080151.1 | NP_001073620.1 | 7, 8 | 1418 | ||
XM_008764155.1 | XP_008762377.1 | 7, 8 | 1499 | |||
XM_008764156.1 | XP_008762378.1 | 7, 8 | 1493 | |||
XM_008764157.1 | XP_008762379.1 | 5, 6 | 1630 | |||
GenBank mRNA | BC129108.1 | - | 7, 8 | 1418 | 7104019 |
