siRNA Details
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025738.1 | NP_001020909.1 | 4 | 432 | ||
XM_006239220.2 | XP_006239282.1 | 4 | 475 | |||
XM_006239221.2 | XP_006239283.1 | 4 | 475 | |||
XM_006239222.2 | XP_006239284.1 | 4 | 473 | |||
XM_006239223.2 | XP_006239285.1 | 4 | 474 | |||
GenBank mRNA | BC098831.1 | - | 4 | 432 | 7378554 | |
FQ227288.1 | - | 4 | 453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
