siRNA Details
Gene Symbol: Rpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025739.1 | NP_001020910.1 | 2, 3 | 601 | ||
XM_006239224.2 | XP_006239286.1 | 3, 4 | 344 | |||
GenBank mRNA | BC097372.1 | - | 2, 3 | 601 | 7444187 | |
FQ217185.1 | - | 3, 4 | 281 | |||
FQ218104.1 | - | 3, 4 | 286 | |||
FQ221293.1 | - | 3, 4 | 285 | |||
FQ221469.1 | - | 3, 4 | 285 | |||
FQ222336.1 | - | 3, 4 | 285 | |||
FQ222758.1 | - | 3, 4 | 285 | |||
FQ222952.1 | - | 3, 4 | 281 | |||
FQ224024.1 | - | 3, 4 | 283 | |||
X62146.1 | - | 3, 4 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
