siRNA Details
Gene Symbol: C1qc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008524.1 | NP_001008524.1 | 3 | 360 | ||
XM_006239225.1 | XP_006239287.1 | 3 | 640 | |||
GenBank mRNA | BC086409.1 | - | 3 | 360 | 7306579 | |
FQ218827.1 | - | 3 | 384 | |||
FQ219610.1 | - | 3 | 412 | |||
FQ228978.1 | - | 3 | 644 |
