siRNA Details
Gene Symbol: Usp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198785.1 | NP_942080.1 | 13 | 1880 | ||
XM_006239234.2 | XP_006239296.1 | 14 | 1914 | |||
XM_006239235.2 | XP_006239297.1 | 14 | 1914 | |||
XM_006239236.2 | XP_006239298.1 | 14 | 1911 | |||
XM_006239237.2 | XP_006239299.1 | 14 | 1911 | |||
GenBank mRNA | AB073880.1 | - | 13 | 1880 |
