siRNA Details
Gene Symbol: Cda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108688.1 | NP_001102158.1 | 353 |
Gene Symbol: LOC100909857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750066.3 | XP_003750114.1 | 3, 4 | 370 |

More Information
Other Species Targeted

Gene Ontology Categories:
