siRNA Details
Gene Symbol: Minos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173556.1 | NP_001167027.1 | 3 | 243 | ||
GenBank mRNA | BC166932.1 | - | 3 | 214 | 6921482 | |
FM054041.1 | - | 3 | 243 | |||
FQ221358.1 | - | 3 | 171 | |||
FQ228668.1 | - | 3 | 245 |
Gene Symbol: LOC100361944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240216.2 | XP_006240278.1 | 1 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
