siRNA Details
Gene Symbol: Emc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108690.1 | NP_001102160.1 | 17, 18 | 2093 | ||
XM_006239245.2 | XP_006239307.1 | 18, 19 | 2124 | |||
GenBank mRNA | FQ216974.1 | - | 18, 19 | 2083 | ||
FQ218240.1 | - | 2, 3 | 248 |
