siRNA Details
Gene Symbol: Arhgef19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108692.1 | NP_001102162.1 | 5 | 988 | ||
XM_008764270.1 | XP_008762492.1 | 3 | 575 | |||
XM_008764271.1 | XP_008762493.1 | 3 | 535 | |||
XM_008764272.1 | XP_008762494.1 | 3 | 1814 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
