siRNA Details
Gene Symbol: Fblim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007554.1 | NP_001007555.1 | 8 | 1107 | ||
XM_008764273.1 | XP_008762495.1 | 8 | 1874 | |||
XM_008764274.1 | XP_008762496.1 | 7 | 1037 | |||
XM_008764275.1 | XP_008762497.1 | 8 | 1106 | |||
XM_008764276.1 | XP_008762498.1 | 8 | 1257 | |||
GenBank mRNA | BC086398.1 | - | 8 | 1107 | 7303205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
