siRNA Details
Gene Symbol: Mthfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239413.2 | XP_006239475.1 | 5 | 2280 | ||
XM_006239414.2 | XP_006239476.1 | 5 | 778 | |||
XM_006239416.1 | XP_006239478.1 | 5 | 945 | |||
XM_006239417.1 | XP_006239479.1 | 5 | 942 | |||
XM_006239418.1 | XP_006239480.1 | 4 | 877 | |||
XM_342975.6 | XP_342976.3 | 5 | 2283 | |||
GenBank mRNA | U57049.1 | - | 4 | 671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
