siRNA Details
Gene Symbol: Dnajc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108694.2 | NP_001102164.1 | 7 | 681 | ||
GenBank mRNA | BC161936.1 | - | 7 | 681 | 9022755 |

More Information
Other Species Targeted

