siRNA Details
Gene Symbol: RGD1304567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009711.1 | NP_001009711.1 | 3 | 237 | ||
GenBank mRNA | BC087640.1 | - | 3 | 237 | 7308844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
