siRNA Details
Gene Symbol: Capn13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025133.1 | NP_001020304.1 | 11 | 1217 | ||
XM_006239777.2 | XP_006239839.1 | 12 | 1375 | |||
XM_006239778.2 | XP_006239840.1 | 11 | 1328 | |||
XM_006239779.2 | XP_006239841.1 | 12 | 1371 | |||
GenBank mRNA | BC091250.1 | - | 11 | 1217 | 7368359 |
