siRNA Details
Gene Symbol: Wdr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037791.2 | NP_001032880.2 | 19 | 2022 | ||
XM_006239770.2 | XP_006239832.1 | 19 | 2030 | |||
GenBank mRNA | BC099168.1 | - | 4 | 521 | ||
BC169117.1 | - | 20 | 2219 |
