siRNA Details
Gene Symbol: Wdr43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037791.2 | NP_001032880.2 | 16 | 1687 | ||
XM_006239770.2 | XP_006239832.1 | 16 | 1695 | |||
GenBank mRNA | BC099168.1 | - | 1 | 186 | ||
BC169117.1 | - | 17 | 1884 |
