siRNA Details
Gene Symbol: Rbks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108703.1 | NP_001102173.1 | 3 | 286 | ||
XM_006239805.2 | XP_006239867.1 | 3 | 332 | |||
XM_006239806.2 | XP_006239868.1 | 3 | 332 | |||
XM_008764520.1 | XP_008762742.1 | 3 | 780 | |||
XM_008764521.1 | XP_008762743.1 | 3 | 332 | |||
GenBank mRNA | FQ209797.1 | - | 3 | 268 |
