siRNA Details
Gene Symbol: Drc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007009.1 | NP_001007010.1 | 11 | 1499 | ||
GenBank mRNA | BC083825.1 | - | 11 | 1499 | 7112171 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Drc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007009.1 | NP_001007010.1 | 11 | 1499 | ||
GenBank mRNA | BC083825.1 | - | 11 | 1499 | 7112171 |