siRNA Details
Gene Symbol: Snx13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108708.1 | NP_001102178.1 | 9, 10 | 1039 | ||
XM_006240022.2 | XP_006240084.1 | 9, 10 | 1506 | |||
XM_006240023.2 | XP_006240085.1 | 9, 10 | 1504 |

More Information
Other Species Targeted

Gene Ontology Categories:
