siRNA Details
Gene Symbol: Etv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108709.3 | NP_001102179.2 | 10 | 1162 | ||
NM_001163156.3 | NP_001156628.1 | 11 | 1250 | |||
XM_006240047.2 | XP_006240109.1 | 10 | 1212 | |||
XM_006240048.2 | XP_006240110.1 | 11 | 1270 | |||
XM_006240050.2 | XP_006240112.1 | 8 | 1356 | |||
XM_008764647.1 | XP_008762869.1 | 9 | 1143 | |||
XM_008764648.1 | XP_008762870.1 | 10 | 1616 | |||
GenBank mRNA | BC168860.1 | - | 10 | 1162 | 7121808 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001163154, NM_007960, AK035914, BC005645, CK619331, and others.
Gene Ontology Categories:
