siRNA Details
Gene Symbol: Etv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108709.3 | NP_001102179.2 | 6 | 444 | ||
NM_001163156.3 | NP_001156628.1 | 6 | 463 | |||
XM_006240047.2 | XP_006240109.1 | 5 | 425 | |||
XM_006240048.2 | XP_006240110.1 | 6 | 483 | |||
XM_006240050.2 | XP_006240112.1 | 3 | 569 | |||
XM_008764647.1 | XP_008762869.1 | 5 | 425 | |||
XM_008764648.1 | XP_008762870.1 | 6 | 898 | |||
GenBank mRNA | BC168860.1 | - | 6 | 444 | 7121808 | |
CB547717.1 | - | 6 | 462 |
