siRNA Details
Gene Symbol: Snx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108711.1 | NP_001102181.1 | 13 | 1190 | ||
GenBank mRNA | BC168856.1 | - | 12 | 1151 | 7458822 |

More Information
Other Species Targeted

Gene Ontology Categories:
