siRNA Details
Gene Symbol: Ppp2r3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014196.1 | NP_001014218.1 | 8 | 827 | ||
XM_008764660.1 | XP_008762882.1 | 7 | 713 | |||
GenBank mRNA | BC079257.1 | - | 8 | 827 | 7133341 | |
FQ227548.1 | - | 7 | 684 |
