siRNA Details
Gene Symbol: Atp5s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007749.1 | NP_001007750.1 | 4 | 650 | ||
XM_006240174.2 | XP_006240236.1 | 4 | 553 | |||
XM_006240175.2 | XP_006240237.1 | 4 | 442 | |||
XM_006240176.2 | XP_006240238.1 | 4 | 392 | |||
GenBank mRNA | BC083655.1 | - | 4 | 650 | 7135201 |
