siRNA Details
Gene Symbol: Trmt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108713.1 | NP_001102183.1 | 4 | 800 | ||
XM_006240213.2 | XP_006240275.1 | 3 | 776 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029580, BC012521, AK220389, AK049127, AK011576.
Gene Ontology Categories:
