siRNA Details
Gene Symbol: Dcaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108716.1 | NP_001102186.1 | 6 | 608 | ||
XM_006240347.2 | XP_006240409.1 | 6 | 608 | |||
XM_008764814.1 | XP_008763036.1 | 5 | 656 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
