siRNA Details
Gene Symbol: RGD1311756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173557.1 | NP_001167028.1 | 9 | 1467 | ||
XM_006240439.2 | XP_006240501.1 | 10 | 1641 | |||
XM_006240441.2 | XP_006240503.1 | 10 | 1702 | |||
XM_006240442.2 | XP_006240504.1 | 8 | 2454 | |||
XM_008764817.1 | XP_008763039.1 | 10 | 1696 | |||
GenBank mRNA | FQ221290.1 | - | 8 | 1323 |
