siRNA Details
Gene Symbol: RGD1311756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173557.1 | NP_001167028.1 | 9, 10 | 1508 | ||
XM_006240439.2 | XP_006240501.1 | 10, 11 | 1682 | |||
XM_006240441.2 | XP_006240503.1 | 10, 11 | 1743 | |||
XM_006240442.2 | XP_006240504.1 | 8, 9 | 2495 | |||
XM_008764817.1 | XP_008763039.1 | 10, 11 | 1737 | |||
GenBank mRNA | FQ221290.1 | - | 8, 9 | 1364 |
