siRNA Details
Gene Symbol: RGD1311756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173557.1 | NP_001167028.1 | 5 | 686 | ||
XM_006240439.2 | XP_006240501.1 | 6 | 860 | |||
XM_006240441.2 | XP_006240503.1 | 6 | 921 | |||
XM_006240442.2 | XP_006240504.1 | 4 | 1673 | |||
XM_008764817.1 | XP_008763039.1 | 6 | 915 | |||
GenBank mRNA | FQ221290.1 | - | 4 | 542 |
