siRNA Details
Gene Symbol: Vrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012194.1 | NP_001012194.1 | 6 | 768 | ||
XM_006240530.2 | XP_006240592.1 | 6 | 594 | |||
XM_006240531.2 | XP_006240593.1 | 6 | 624 | |||
XM_006240532.2 | XP_006240594.1 | 6 | 1016 | |||
XM_006240533.2 | XP_006240595.1 | 6 | 503 | |||
XM_008764822.1 | XP_008763044.1 | 6 | 1016 | |||
GenBank mRNA | BC079130.1 | - | 6 | 768 | 7127355 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011705, NM_001029844, NM_001029843, AK088674, BC016676, and others.
Gene Ontology Categories:
