siRNA Details
Gene Symbol: Rapgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047915.1 | NP_001041380.1 | 4, 5 | 701 | ||
GenBank mRNA | AY390379.1 | - | 4, 5 | 701 | ||
FQ211964.1 | - | 6, 7 | 921 |

More Information
Other Species Targeted

Gene Ontology Categories:
