siRNA Details
Gene Symbol: Rnf41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012195.1 | NP_001012195.1 | 6 | 789 | ||
XM_006240772.2 | XP_006240834.1 | 8 | 1220 | |||
XM_006240773.2 | XP_006240835.1 | 8 | 1282 | |||
XM_006240774.2 | XP_006240836.1 | 7 | 1128 | |||
XM_006240775.2 | XP_006240837.1 | 8 | 1138 | |||
XM_006240776.2 | XP_006240838.1 | 7 | 1046 | |||
XM_008765030.1 | XP_008763252.1 | 7 | 2139 | |||
GenBank mRNA | BC083614.1 | - | 6 | 789 | 7133774 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
