siRNA Details
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033070.1 | NP_001028242.1 | 5 | 425 | ||
XM_006240781.2 | XP_006240843.1 | 5 | 454 | |||
XM_008765031.1 | XP_008763253.1 | 5 | 449 | |||
GenBank mRNA | BC100070.1 | - | 5 | 425 | 7385206 |
