siRNA Details
Gene Symbol: Sarnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033070.1 | NP_001028242.1 | 8, 9 | 598 | ||
XM_008765031.1 | XP_008763253.1 | 8, 9 | 622 | |||
GenBank mRNA | BC100070.1 | - | 8, 9 | 598 | 7385206 |

More Information
Other Species Targeted

Gene Ontology Categories:
