siRNA Details
Gene Symbol: Mfsd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108730.1 | NP_001102200.1 | 8 | 1243 | ||
XM_008765107.1 | XP_008763329.1 | 7 | 1140 |

More Information
Other Species Targeted

