siRNA Details
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040190.1 | NP_001035280.1 | 7 | 1167 | ||
GenBank mRNA | AY611623.1 | - | 7 | 1167 | ||
BC128695.1 | - | 7 | 1019 | 8366437 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178662, AK028298, AK039411, AK038483, AK086723, and others.
Gene Ontology Categories:
