siRNA Details
Gene Symbol: Atcay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040190.1 | NP_001035280.1 | 7 | 1246 | ||
GenBank mRNA | AY611623.1 | - | 7 | 1246 | ||
BC128695.1 | - | 7 | 1098 | 8366437 |

More Information
Other Species Targeted

Gene Ontology Categories:
